
Benalaxyl-M

Synonym(s):
N-(2,6-Dimethylphenyl)-N-(phenylacetyl)-D-alanine methyl ester
Empirical Formula (Hill Notation):
C20H23NO3
CAS Number:
Molecular Weight:
325.40
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing