
Atazanavir for system suitability

Synonym(s):
Atazanavir, 1,14-Dimethyl (3S,8S,9S,12S)-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate, BMS-232632
Empirical Formula (Hill Notation):
C38H52N6O7
CAS Number:
Molecular Weight:
704.86
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing