
5α-Dihydrotestosterone-D3 (16,16,17-D3) solution

Synonym(s):
5α-DHT-D3 (16, 16, 17-D3) solution
Empirical Formula (Hill Notation):
C19H27O2D3
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing