Skip to Content
Merck
3-Methoxybenzyl chloride 97%

3-Methoxybenzyl chloride

3-Methoxybenzyl chloride 97%
Synonym(s):
3-(Chloromethyl)anisole
Linear Formula:
CH3OC6H4CH2Cl
CAS Number:
Molecular Weight:
156.61

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing