
1,2-Dipalmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

Synonym(s):
1,2-Dihexadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dipalmitoyl, DPPG-Na, Hexadecanoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, PG(16:0/16:0)
Linear Formula:
C38H74O10PNa
CAS Number:
Molecular Weight:
744.95
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing