
1,2-Dilinoleoyl-sn-glycero-3-phosphocholine

Synonym(s):
(7R,18Z,21Z)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[[(9Z,12Z)-1-oxo-9,12-octadecadien-1-yl]oxy]-3,5,9-trioxa-4-phosphaheptacosa-18,21-dien-1-aminium 4-oxide, inner salt, 1,2-Di(cis-9,12-octadecadienoyl)-sn-glycero-3-phosphocholine, 1,2-Dilinoleoyl-sn-3-glycerophosphorylcholine, 3-sn-Phosphatidylcholine, 1,2-dilinoleoyl, L-α-Dilinoleoyl phosphatidylcholine, L-Dilinoleoyllecithin, Dilinoleoyl-L-α-phosphatidylcholine, PC(18:2(9Z,12Z)/18:2(9Z,12Z))
Empirical Formula (Hill Notation):
C44H80NO8P
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing