Skip to Content
Merck
All Photos(1)

Key Documents

HTS015RTA

Sigma-Aldrich

Ready-to-Assay CX3CR1 Chemokine Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

Ready-to-Assay CX3CR1 Chemokine Receptor Frozen Cells, Human CX3CR1 GPCR frozen cells for Calcium Flux FLIPR Assays.

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

UniProt accession no.

detection method

fluorometric

shipped in

dry ice

General description

Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

CX3CR1 is a GPCR expressed on natural killer cells, cytotoxic T lymphocytes, and macrophages. The sole ligand for CX3CR1, fractalkine, is an unusual chemokine that is expressed as a transmembrane molecule with a CX3C domain and a mucin domain (Imai et al., 1997). Fractalkine is highly expressed on endothelial cells activated by TNFα and other proinflammatory cytokines, and fractalkine/CX3CR1 interactions mediate recruitment of macrophages into the atherosclerotic plaque (Lesnick et al., 2003; McDermott et al., 2003). In addition, fractalkine and CX3CR1 have been implicated in the pathogenesis of glomerulonephritis, HIV infection and rheumatoid arthritis (Ito et al., 2002; Faure et al., 2003; Nanki et al., 2002). Millipore′s cloned human CX3CR1 –expressing cell line is made in the Chem-4 host, which supports high levels of recombinant CX3CR1 expression on the cell surface and contains optimized levels of a recombinant promiscuous G protein to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at CX3CR1.

Cell Line Description

Chem-4
GPCR Cell Lines

Application

Human CX3CR1 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

GPCR Class: A
Protein Target: CX3CR1
Target Sub-Family: Chemokine

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.

Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service