Skip to Content
Merck
All Photos(1)

Key Documents

676504

Sigma-Aldrich

VEGFR Tyrosine Kinase Inhibitor VI, AAL-993

The VEGFR Tyrosine Kinse Inhibitor VI, ALL-993, also referenced under CAS 269390-77-4, controls the biological activity of VEGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

VEGFR Tyrosine Kinase Inhibitor VI, AAL-993, 2-((4-Pyridyl)methyl)amino-N-(3-(trifluoromethyl)phenyl)benzamide, ZK260253, PDGFR Tyrosine Kinase Inhibitor XXIII, VEGFR2 Kinase Inhibitor XXXIII, VEGFR1 Kinase Inhibitor VIII, VEGFR3 Kinase Inhibitor VII, AAL993, PDGFR Tyrosine Kinase Inhibitor XXIII, 2-((4-Pyridyl)methyl)amino-N-(3-(trifluoromethyl)phenyl)benzamide, ZK260253, VEGFR2 Kinase Inhibitor XXXIII, VEGFR1 Kinase Inhibitor VIII, VEGFR3 Kinase Inhibitor VII, AAL993

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16F3N3O
CAS Number:
Molecular Weight:
371.36
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H16F3N3O/c21-20(22,23)15-4-3-5-16(12-15)26-19(27)17-6-1-2-7-18(17)25-13-14-8-10-24-11-9-14/h1-12,25H,13H2,(H,26,27)

InChI key

BLAFVGLBBOPRLP-UHFFFAOYSA-N

General description

A cell-permeable anthranilamide that acts as a potent VEGFR inhibitor (IC50 = 130, 23, and 18 nM against VEGFR-1, -2, and -3, respectively; IC50 = 1.24 nM against VEGF-induced human VEGFR-2 phosphorylation in CHO cells) by targeting the ATP-binding site of VEGFR in its inactive "DFG-out" conformation and effectively suppresses tumor growths (50 to 100 mg/kg; p.o.) via its anti-angiogenesis activity in mice and rats in vivo, while inhibiting c-kit, CSF-1R/c-Fms, PDGFR-β, and c-Abl only at higher concentrations (IC50 = 236, 380, 640, and 2820 nM, respectively). AAL933 and two other VEGFR inhibitors, KRN633 and SU5416, are also shown to inhibit hypoxia-induced HIF-1α expression (by <90% at 30 M) and transcription activation. Unlike KRN633 and SU5416, AAL933 prevents only hypoxia-induced Erk, but not Akt, phosphorylation in HeLa cells.
A cell-permeable anthranilamide that acts as a potent VEGFR inhibitor (IC50 = 130, 23, and 18 nM against VEGFR-1, -2, and -3, respectively; IC50 = 1.24 nM against VEGF-induced human VEGFR-2 phosphorylation in CHO cells) by targeting the ATP-binding site of VEGFR in its inactive "DFG-out" conformation, while inhibiting c-kit, CSF-1R/c-Fms, PDGFR-β, and c-Abl only at higher concentrations (IC50 = 236, 380, 640, and 2820 nM, respectively) and displaying little or no activity against EGFR/HER-1/ErbB (IC50 = 10.4 M) and six other kinases (IC50 ≥10 M). Reported to be orally available in mice (Conc. in plasma = 9.5, 4.7, 1.5, and 1.1 M, respectively, 0.5 h, 1 h, 1.5 h, and 2 h after p.o. dosage of 50 mg/kg), rats, and dogs, and effectively suppress tumor growths (50 to 100 mg/kg; p.o.) via its anti-angiogenesis activity in various cancer models in vivo, including murine melanoma B16-BL6, rat mammary adenocarcinoma BN472, as well as human prostate cancer DU145-grafted mice in vivo. AAL933 and two other VEGFR inhibitors, KRN633 and SU5416, are also shown to inhibit hypoxia- (1% O2) induced HIF-1α expression (by <90% at 30 M) and transcription activation (IC50 = 6.53, 3.79, and 8.24 M, respectively, by AAL933, KRN633, and SU5416 in HeLa-HRE-Luc reporter assays). However, unlike KRN633 and SU5416, AAL933 prevents only hypoxia-induced Erk, but not Akt, phosphorylation in HeLa cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ban, H.S., et al. 2010. Cancer Lett.296, 17; Manley, P.W., et al. 2004. Biochim. Biophys. Acta1697, 17; Manley, P.W., et al. 2002. J. Med. Chem.45, 5687.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno da Rocha-Azevedo et al.
Cell reports, 32(13), 108187-108187 (2020-10-01)
The dynamic nanoscale organization of cell surface receptors plays an important role in signaling. We determine this organization and its relation to activation of VEGF receptor-2 (VEGFR-2), a critical receptor tyrosine kinase in endothelial cells (ECs), by combining single-molecule imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service