GF70512331
Tungsten-copper alloy, W60Cu40
175mm tube, 0.5mm outside diameter, 0.13mm wall thickness, 0.24mm inside diameter
Synonym(s):
W 187100
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 705-123-31
L × wall thickness
175 mm × 0.13 mm
O.D. × I.D.
0.5 mm × 0.24 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service