BMSH3770SEUK
Benchmark Magnetic 7 x 7 inch Stirrer
AC/DC input 230 V, UK plug
Synonym(s):
stirrer, stirring, magnetic Stirrers, , ceramic top stirrer,
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
ceramic platform
manufacturer/tradename
Benchmark Scientific H3770-S-E-UK
parameter
150-1500 rpm speed
plate W × L
18 cm × 18 cm
Looking for similar products? Visit Product Comparison Guide
General description
Benchmark Scientific Hotplates and Stirrers provide reliable performance and safe operation for everyday laboratory use. Our space efficient 7 x 7inch hotplate/stirrer series features a durable, chemical resistant white ceramic top plate and advanced microprocessor controls. The constant monitoring system provides precise control of temperature with rapid heating and faster boiling times. In addition, the powerful motor drive system ensures efficient mixing in larger and more viscous samples. Temperature and mixing speed can be viewed on the large digital display and a safety LED illuminates when the top plate temperature exceeds 50°C. An external temperature probe can also be used for applications that require precise control of sample temperature. The probe provides direct feedback to the microprocessor with constant monitoring and adjustments for maintaining the selected temperature in the sample within +/ 1°C. All models are compatible with borosilicate glass beakers and other vessels with up to 5 L maximum stirring volume. An optional clamp assembly (H3770CS) is available for mounting thermometers, temperature probes, etc.
Application
Stirring of liquids in various vessels, up to 5L
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service