Merck
  • Development of hepatoma-derived, bidirectional oval-like cells as a model to study host interactions with hepatitis C virus during differentiation.
Development of hepatoma-derived, bidirectional oval-like cells as a model to study host interactions with hepatitis C virus during differentiation.
Oncotarget (2017-09-15)
Masahiko Ito, Suofeng Sun, Takasuke Fukuhara, Ryosuke Suzuki, Miho Tamai, Toyohiko Yamauchi, Kenji Nakashima, Yoh-Ichi Tagawa, Shigetoshi Okazaki, Yoshiharu Matsuura, Takaji Wakita, Tetsuro Suzuki
ABSTRACT
Directed differentiation of human stem cells including induced pluripotent stem cells into hepatic cells potentially leads to acquired susceptibility to hepatitis C virus (HCV) infection. However, cellular determinants that change their expression during cell reprogramming or hepatic differentiation and are pivotal for supporting the HCV life cycle remain unclear. In this study, by introducing a set of reprogramming factors, we established HuH-7-derived oval-like cell lines, Hdo-17 and -23, which possess features of bipotential liver precursors. Upon induction of hepatocyte differentiation, expression of mature hepatocyte markers and hepatoblast markers in cells increased and decreased, respectively. In contrast, in response to cholangiocytic differentiation induction, gene expression of epithelium markers increased and cells formed round cysts with a central luminal space. Hdo cells lost their susceptibility to HCV infection and viral RNA replication. Hepatic differentiation of Hdo cells potentially led to recovery of permissiveness to HCV RNA replication. Gene expression profiling showed that most host-cell factors known to be involved in the HCV life cycle, except CD81, are expressed in Hdo cells comparable to HuH-7 cells. HCV pseudoparticle infectivity was significantly but partially recovered by ectopic expression of CD81, suggesting possible involvement of additional unidentified factors in HCV entry. In addition, we identified miR200a-3p, which is highly expressed in Hdo cells and stem cells but poorly expressed in differentiated cells and mature hepatocytes, as a novel negative regulator of HCV replication. In conclusion, our results showed that epigenetic reprogramming of human hepatoma cells potentially changes their permissivity to HCV.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-LDLR Antibody, clone C7, clone C7, from mouse
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)