Merck
  • A ratiometric fluorescence probe for imaging sulfur dioxide derivatives in the mitochondria of living cells.
A ratiometric fluorescence probe for imaging sulfur dioxide derivatives in the mitochondria of living cells.
Organic & biomolecular chemistry (2017-01-18)
Yongfei Wang, Qingtao Meng, Run Zhang, Hongmin Jia, Xuehu Zhang, Zhiqiang Zhang
ABSTRACT
Although sulfur dioxide (SO
Sigma-Aldrich
Iodoethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
6-hydroxy-2-naphthaldehyde, AldrichCPR
Sigma-Aldrich
1,3-Propylene sulfite, 99%