Merck
  • A new class of highly potent, broadly neutralizing antibodies isolated from viremic patients infected with dengue virus.
A new class of highly potent, broadly neutralizing antibodies isolated from viremic patients infected with dengue virus.
Nature immunology (2014-12-17)
Wanwisa Dejnirattisai, Wiyada Wongwiwat, Sunpetchuda Supasa, Xiaokang Zhang, Xinghong Dai, Alexander Rouvinski, Amonrat Jumnainsong, Carolyn Edwards, Nguyen Than Ha Quyen, Thaneeya Duangchinda, Jonathan M Grimes, Wen-Yang Tsai, Chih-Yun Lai, Wei-Kung Wang, Prida Malasit, Jeremy Farrar, Cameron P Simmons, Z Hong Zhou, Felix A Rey, Juthathip Mongkolsapaya, Gavin R Screaton
ABSTRACT
Dengue is a rapidly emerging, mosquito-borne viral infection, with an estimated 400 million infections occurring annually. To gain insight into dengue immunity, we characterized 145 human monoclonal antibodies (mAbs) and identified a previously unknown epitope, the envelope dimer epitope (EDE), that bridges two envelope protein subunits that make up the 90 repeating dimers on the mature virion. The mAbs to EDE were broadly reactive across the dengue serocomplex and fully neutralized virus produced in either insect cells or primary human cells, with 50% neutralization in the low picomolar range. Our results provide a path to a subunit vaccine against dengue virus and have implications for the design and monitoring of future vaccine trials in which the induction of antibody to the EDE should be prioritized.
Sigma-Aldrich
Anti-Human IgM (μ-chain specific)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)-Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution