Merck
[Biological functions of allantoin].
Izvestiia Akademii nauk. Seriia biologicheskaia (2006-11-08)
A V Shestopalov, T P Shkurat, Z I Mikashinovich, I O Kryzhanovskaia, M A Bogacheva, S V Lomteva, V P Prokof'ev, E P Gus'kov
ABSTRACT
A historical review of allantoin research is presented. An increased allantoin level in the trophoblast and serum of pregnant women has been demonstrated. Allantoin concentration decreased in placental tissues and increased in the serum in developing placental insufficiency.
Product Description
Allantoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Allantoin, analytical standard
Sigma-Aldrich
Allantoin, ≥98.0% (N)
Supelco
Allantoin, Pharmaceutical Secondary Standard; Certified Reference Material