Merck
  • A new phenolic compound, 4-dehydrochebulic acid-1,6-dimethyl ester from Sapium insigne leaves.
A new phenolic compound, 4-dehydrochebulic acid-1,6-dimethyl ester from Sapium insigne leaves.
Journal of natural medicines (2009-12-29)
Hari Prasad Devkota, Purusotam Basnet, Shoji Yahara
ABSTRACT
A new phenolic compound, 4-dehydrochebulic acid-1,6-dimethyl ester (2) was isolated from the leaves of Sapium insigne (Royle) Benth. ex Hook. fil. (Euphorbiaceae) along with three known compounds gallic acid (1), brevifolin carboxylic acid (3) and fraxin (4). Structures of those compounds were elucidated on the basis of spectroscopic data.
Sigma-Aldrich
Chebulinic acid, ≥85% (LC/MS-ELSD)