Merck
  • 2-Acyl-tetrahydroisoquinoline-3-carboxylic acids: lead compounds with triple actions, peroxisome proliferator-activated receptor α/γ agonist and protein-tyrosine phosphatase 1B inhibitory activities.
2-Acyl-tetrahydroisoquinoline-3-carboxylic acids: lead compounds with triple actions, peroxisome proliferator-activated receptor α/γ agonist and protein-tyrosine phosphatase 1B inhibitory activities.
Chemical & pharmaceutical bulletin (2011-07-02)
Kazuya Otake, Satoru Azukizawa, Kenji Takahashi, Masaki Fukui, Michiko Shibabayashi, Hikaru Kamemoto, Masayasu Kasai, Hiroaki Shirahase
ABSTRACT
2-Acyl-tetrahydroisoquinoline-3-carboxylic acid derivatives were synthesized and biologically evaluated. (S)-2-(2,4-Hexadienoyl)-7-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (14) showed peroxisome proliferator-activated receptor γ (PPARγ) and PPARα agonist activities and protein-tyrosine phosphatase 1B (PTP-1B) inhibitory activities. PPARγ agonist activity of 14 was comparable to that of rosiglitazone, and PTP-1B inhibitory activity was about 10-fold weaker than that of ertiprotafib, a PTP-1B inhibitor. Compound 14 showed high oral absorption in rats and potent hypoglycemic effects in KK-A(y) mice. In conclusion, 14 would be an excellent lead compound for a new type of anti-diabetic drug with triple actions.
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%