Merck
  • Design, synthesis, and evaluation of stable and taste-free erythromycin proprodrugs.
Design, synthesis, and evaluation of stable and taste-free erythromycin proprodrugs.
Journal of medicinal chemistry (2005-05-27)
Pranab K Bhadra, Gareth A Morris, Jill Barber
ABSTRACT
Erythromycin A is normally formulated for children as its 2'-ethyl succinate, a taste-free prodrug. Unfortunately, the prodrug hydrolyzes at a measurable rate in the medicine bottle, leading to the vile-tasting erythromycin. We have prepared derivatives of erythromycin B as putative paediatric prodrugs, taking advantage of the much improved acid stability of erythromycin B relative to erythromycin A. Thus, erythromycin B enol ether ethyl succinate is very poorly soluble in water, and its hydrolysis is undetectable in conditions resembling the medicine bottle. In acid, however, it converts rapidly to erythromycin B 2'-ethyl succinate, and this is in turn hydrolyzed to erythromycin B in neutral and basic conditions. Derivatives of erythromycin B enol ether are therefore proposed as taste-free proprodrugs of erythromycin B.
Product Description
Erythromycin B, European Pharmacopoeia (EP) Reference Standard