Merck
  • Avasimibe, an ACAT inhibitor, enhances the lipid lowering effect of atorvastatin in subjects with homozygous familial hypercholesterolemia.
Avasimibe, an ACAT inhibitor, enhances the lipid lowering effect of atorvastatin in subjects with homozygous familial hypercholesterolemia.
Atherosclerosis (2003-12-04)
Frederick J Raal, A David Marais, Ellen Klepack, Jennifer Lovalvo, Richard McLain, Therese Heinonen
ABSTRACT
This study assessed the efficacy and safety of avasimibe (CI-1011), an inhibitor of acyl coenzyme A-cholesterol acyltransferase (ACAT) in subjects with homozygous familial hypercholesterolemia (HoFH). Twenty seven subjects were enrolled in a double-blind, randomized, 3-sequence crossover trial of atorvastatin 80 mg QD, avasimibe 750 mg QD, and the combined treatment of atorvastatin 80 mg QD and avasimibe 750 mg QD after a washout period of 4 weeks. Each treatment period was administered over 6 weeks for a total of 18 weeks. There were no significant lipid changes resulting from the administration of avasimibe monotherapy. Avasimibe in combination with atorvastatin resulted in a significantly better reduction of total cholesterol (TC) as compared to atorvastatin alone (-22% versus -18%) (P < 0.05). All other lipid changes were not statistically significant for combination therapy compared to atorvastatin monotherapy, however there were greater reductions in triglycerides (TG) (-24% versus -13%), low-density lipoprotein cholesterol (LDL-C) (-23% versus -19%), very low-density lipoprotein cholesterol (VLDL-C) (-24% versus -13%) and high-density lipoprotein cholesterol (HDL-C) (-11% versus -6%). Avasimibe may modestly enhance the lipid-reducing effect of atorvastatin by further inhibiting the production of intracellular cholesterol through mechanisms that appear to be compatible in this population.
Sigma-Aldrich
Avasimibe, ≥98% (HPLC)
Pricing and availability is not currently available.