Merck
Topical protection against long-wave ultraviolet A.
Journal of the American Academy of Dermatology (1983-09-01)
M Jarratt, M Hill, K Smiles
ABSTRACT
A PABA ester-oxybenzone preparation is superior to PABA or sulisobenzone alone in protecting the skin from methoxsalen-induced ultraviolet A (UVA) phototoxicity after water substantivity challenge. Such a mixture would be useful as a UVA screen for uninvolved or actinically damaged skin in patients receiving psoralens and ultraviolet A (PUVA) therapy. An effective topical UVA screen also may protect against UVA-induced diseases like solar urticaria, polymorphic light eruptions, drug-induced phototoxicity or photoallergy, and possibly against the deep degenerative changes of solar elastosis.
Sigma-Aldrich
5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, ≥97.0% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Supelco
Sulisobenzone, analytical standard
Pricing and availability is not currently available. Contact Technical Service