Merck
  • Mechanistic and conformational studies on the interaction of food dye amaranth with human serum albumin by multispectroscopic methods.
Mechanistic and conformational studies on the interaction of food dye amaranth with human serum albumin by multispectroscopic methods.
Food chemistry (2012-11-06)
Guowen Zhang, Yadi Ma
ABSTRACT
The mechanism of interaction between food dye amaranth and human serum albumin (HSA) in physiological buffer (pH 7.4) was investigated by fluorescence, UV-vis absorption, circular dichroism (CD), and Fourier transform infrared (FT-IR) spectroscopy. Results obtained from analysis of fluorescence spectra indicated that amaranth had a strong ability to quench the intrinsic fluorescence of HSA through a static quenching procedure. The negative value of enthalpy change and positive value of entropy change elucidated that the binding of amaranth to HSA was driven mainly by hydrophobic and hydrogen bonding interactions. The surface hydrophobicity of HSA increased after binding with amaranth. The binding distance between HSA and amaranth was estimated to be 3.03 nm and subdomain IIA (Sudlow site I) was the primary binding site for amaranth on HSA. The results of CD and FT-IR spectra showed that binding of amaranth to HSA induced conformational changes of HSA.
Product Description
Supelco
Amaranth, analytical standard
Sigma-Aldrich
Amaranth, Dye content 85-95 %