Merck
  • Controlled synthesis, growth mechanism and highly efficient solar photocatalysis of nitrogen-doped bismuth subcarbonate hierarchical nanosheets architectures.
Controlled synthesis, growth mechanism and highly efficient solar photocatalysis of nitrogen-doped bismuth subcarbonate hierarchical nanosheets architectures.
Dalton transactions (Cambridge, England : 2003) (2012-05-25)
Fan Dong, Yanjuan Sun, Wing-Kei Ho, Zhongbiao Wu
ABSTRACT
The synthesis and self-assembly of hierarchical architectures from nanoscale building blocks with unique morphology, orientation and dimension have opened up new opportunities to enhance their functional performances and remain a great challenge. This work represents tunable synthesis of various types of 3D monodisperse in situ N-doped (BiO)(2)CO(3) hierarchical architectures composed of 2D single-crystal nanosheets with dominant (001) facets by a one-pot template-free hydrothermal method from bismuth citrate and ammonia solution. Depending on the concentration of ammonia solution, the morphology of N-doped (BiO)(2)CO(3), including dandelion-like, hydrangea-like and peony flower-like microspheres, can be selectively constructed due to different self-assembly patterns of nanosheets. It was revealed that the ammonia played dual roles in the formation of N-doped (BiO)(2)CO(3) architectures. One is to hydrolyze bismuth citrate, and the other is to behave as a nitrogen doping source. The in situ doped nitrogen substituted for oxygen in (BiO)(2)CO(3) and subsequently narrowed the band gap, making N-doped (BiO)(2)CO(3) visible light active. Due to the special nanosheets architectures, the prepared various N-doped (BiO)(2)CO(3) materials exhibited especially efficient photocatalytic activity and high durability for the removal of NO in air under both visible and UV light irradiation. Based on the direct observation of the growth process with respect to phase structure, chemical composition and morphological structure, a novel growth mechanism is revealed, which involves a unique multistep pathway, including reaction-nucleation, aggregation, crystallization, dissolution-recrystallization, and Ostwald ripening. The facile synthesis approach and the proposed growth mechanism could provide new insights into the design and controlled synthesis of inorganic hierarchical materials with new or enhanced properties.
Sigma-Aldrich
Bismuth(III) carbonate basic, puriss., meets analytical specification of Ph. Eur., 80-82.5% Bi basis (calc. on dried substance)