Merck
  • Cationic cyclization of 2-alkenyl-1,3-dithiolanes: diastereoselective synthesis of trans-decalins.
Cationic cyclization of 2-alkenyl-1,3-dithiolanes: diastereoselective synthesis of trans-decalins.
The Journal of organic chemistry (2011-03-31)
Sylvie Goncalves, Stefano Santoro, Marc Nicolas, Alain Wagner, Philippe Maillos, Fahmi Himo, Rachid Baati
ABSTRACT
An unprecedented and highly diastereoselective 6-endo-trig cyclization of 2-alkenyl-1,3-dithiolanes has been developed yielding trans-decalins, an important scaffold present in numerous di- and triterpenes. The novelty of this 6-endo-trig cyclization stands in the stepwise mechanism involving 2-alkenyl-1,3-dithiolane, acting as a novel latent initiator. It is suggested that the thioketal opens temporarily under the influence of TMSOTf, triggering the cationic 6-endo-trig cyclization, and closes after C-C bond formation and diastereoselective protonation to terminate the process. DFT calculations confirm this mechanistic proposal and provide a rationale for the observed diastereoselectivity. The reaction tolerates a wide range of functionalities and nucleophilic partners within the substrate. We have also shown that the one-pot 6-endo-trig cyclization followed by in situ 1,3-dithiolane deprotection afford directly the corresponding ketone. This improvement allowed the achievement of the shortest total synthesis of triptophenolide and the shortest formal synthesis of triptolide.
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%
Sigma-Aldrich
cis-Decahydronaphthalene, 99%
Sigma-Aldrich
trans-Decahydronaphthalene, 99%
Sigma-Aldrich
1,3-Dithiolane, 97%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%