Merck
  • Development of a fully automated sequential injection solid-phase extraction procedure coupled to liquid chromatography to determine free 2-hydroxy-4-methoxybenzophenone and 2-hydroxy-4-methoxybenzophenone-5-sulphonic acid in human urine.
Development of a fully automated sequential injection solid-phase extraction procedure coupled to liquid chromatography to determine free 2-hydroxy-4-methoxybenzophenone and 2-hydroxy-4-methoxybenzophenone-5-sulphonic acid in human urine.
Analytica chimica acta (2010-04-07)
Zacarías León, Alberto Chisvert, Angel Balaguer, Amparo Salvador
ABSTRACT
2-Hydroxy-4-methoxybenzophenone and 2-hydroxy-4-methoxybenzophenone-5-sulphonic acid, commonly known as benzophenone-3 (BZ3) and benzophenone-4 (BZ4), respectively, are substances widely used as UV filters in cosmetic products in order to absorb UV radiation and protect human skin from direct exposure to the deleterious wavelengths of sunlight. As with other UV filters, there is evidence of their percutaneous absorption. This work describes an analytical method developed to determine trace levels of free BZ3 and BZ4 in human urine. The methodology is based on a solid-phase extraction (SPE) procedure for clean-up and pre-concentration, followed by the monitoring of the UV filters by liquid chromatography-ultraviolet spectrophotometry detection (LC-UV). In order to improve not only the sensitivity and selectivity, but also the precision of the method, the principle of sequential injection analysis was used to automate the SPE process and to transfer the eluates from the SPE to the LC system. The application of a six-channel valve as an interface for the switching arrangements successfully allowed the on-line connection of SPE sample processing with LC analysis. The SPE process for BZ3 and BZ4 was performed using octadecyl (C18) and diethylaminopropyl (DEA) modified silica microcolumns, respectively, in which the analytes were retained and eluted selectively. Due to the matrix effects, the determination was based on standard addition quantification and was fully validated. The relative standard deviations of the results were 13% and 6% for BZ3 and BZ4, respectively, whereas the limits of detection were 60 and 30 ng mL(-1), respectively. The method was satisfactorily applied to determine BZ3 and BZ4 in urine from volunteers that had applied a sunscreen cosmetic containing both UV filters.
Sigma-Aldrich
5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, ≥97.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sulisobenzone, analytical standard