Merck
  • Synthesis, characterization, and pharmacokinetic studies of PEGylated glucagon-like peptide-1.
Synthesis, characterization, and pharmacokinetic studies of PEGylated glucagon-like peptide-1.
Bioconjugate chemistry (2005-03-17)
Sang-Heon Lee, Seulki Lee, Yu Seok Youn, Dong Hee Na, Su Young Chae, Youngro Byun, Kang Choon Lee
ABSTRACT
Glucagon-like peptide-1-(7-36) (GLP-1) is a hormone derived from the proglucagon molecule, which is considered a highly desirable antidiabetic agent mainly due to its unique glucose-dependent stimulation of insulin secretion profiles. However, the development of a GLP-1-based pharmaceutical agent has a severe limitation due to its very short half-life in plasma, being primarily degraded by dipeptidyl peptidase IV (DPP-IV) enzyme. To overcome this limitation, in this article we propose a novel and potent DPP-IV-resistant form of a poly(ethylene glycol)-conjugated GLP-1 preparation and its pharmacokinetic evaluation in rats. Two series of mono-PEGylated GLP-1, (i) N-terminally modified PEG(2k)-N(ter)-GLP-1 and (ii) isomers of Lys(26), Lys(34) modified PEG(2k)-Lys-GLP-1, were prepared by using mPEG-aldehyde and mPEG-succinimidyl propionate, respectively. To determine the optimized condition for PEGylation, the reactions were monitored at different pH buffer and time intervals by RP-HPLC and MALDI-TOF-MS. The in vitro insulinotropic effect of PEG(2k)-Lys-GLP-1 showed comparable biological activity with native GLP-1 (P = 0.11) in stimulating insulin secretion in isolated rat pancreatic islet and was significantly more potent than the PEG(2k)-N(ter)-GLP-1 (P < 0.05) that showed a marked reduced potency. Furthermore, PEG(2k)-Lys-GLP-1 was clearly resistant to purified DPP-IV in buffer with 50-fold increased half-life compared to unmodified GLP-1. When PEG(2k)-Lys-GLP-1 was administered intravenously and subcutaneously into rats, PEGylation improved the half-life, which resulted in substantial improvement of the mean plasma residence time as a 16-fold increase for iv and a 3.2-fold increase for sc. These preliminary results suggest a site specifically mono-PEGylated GLP-1 greatly improved the pharmacological profiles; thus, we anticipated that it could serve as potential candidate as an antidiabetic agent for the treatment of non-insulin-dependent diabetes patients.
Sigma-Aldrich
Glucagon-Like Peptide I Amide Fragment 7-36 human, ≥97% (HPLC), powder