Merck
Human Liver Memory CD8+ T Cells Use Autophagy for Tissue Residence.
Cell reports (2020-01-23)
Leo Swadling, Laura J Pallett, Mariana O Diniz, Josephine M Baker, Oliver E Amin, Kerstin A Stegmann, Alice R Burton, Nathalie M Schmidt, Anna Jeffery-Smith, Nekisa Zakeri, Kornelija Suveizdyte, Farid Froghi, Giuseppe Fusai, William M Rosenberg, Brian R Davidson, Anna Schurich, A Katharina Simon, Mala K Maini
ABSTRACT
Tissue-resident memory T cells have critical roles in long-term pathogen and tumor immune surveillance in the liver. We investigate the role of autophagy in equipping human memory T cells to acquire tissue residence and maintain functionality in the immunosuppressive liver environment. By performing ex vivo staining of freshly isolated cells from human liver tissue, we find that an increased rate of basal autophagy is a hallmark of intrahepatic lymphocytes, particularly liver-resident CD8+ T cells. CD8+ T cells with increased autophagy are those best able to proliferate and mediate cytotoxicity and cytokine production. Conversely, blocking autophagy induction results in the accumulation of depolarized mitochondria, a feature of exhausted T cells. Primary hepatic stellate cells or the prototypic hepatic cytokine interleukin (IL)-15 induce autophagy in parallel with tissue-homing/retention markers. Inhibition of T cell autophagy abrogates tissue-residence programming. Thus, upregulation of autophagy adapts CD8+ T cells to combat mitochondrial depolarization, optimize functionality, and acquire tissue residence.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
MRT68921 dihydrochloride, ≥98% (HPLC)