Merck
  • Discordance between imaging and immunohistochemistry in unilateral primary aldosteronism.
Discordance between imaging and immunohistochemistry in unilateral primary aldosteronism.
Clinical endocrinology (2017-08-09)
Aya T Nanba, Kazutaka Nanba, James B Byrd, James J Shields, Thomas J Giordano, Barbara S Miller, William E Rainey, Richard J Auchus, Adina F Turcu
ABSTRACT
Correct subtyping of primary aldosteronism (PA) is essential for good surgical outcomes. Adrenal vein sampling (AVS) and/or computed tomography (CT) are used for PA subclassification. Clinical and/or biochemical improvement after surgery, however, is not always achieved in patients with presumed unilateral PA. We aimed to identify the pitfalls in PA subclassification leading to surgical treatment failures. We retrospectively studied 208 patients who underwent adrenal vein sampling (AVS) for PA subclassification in a tertiary referral centre, between January 2009 and August 2016. Simultaneous bilateral AVS was performed before and after cosyntropin administration. We implemented immunohistochemistry for aldosterone synthase (CYP11B2) and 17α-hydroxylase/17,20 lyase (CYP17A1) in adrenal glands resected from patients without improvement of PA after surgical treatment and from those with limitations in AVS interpretation. Of 55 patients who underwent adrenalectomy, three (5.5%) had no improvement of PA. All three patients underwent partial adrenalectomy to remove a CT-detected nodule present on the same side with AVS lateralization. Immunohistochemistry revealed a CYP11B2-negative nodule in both cases available. All patients who underwent total adrenalectomy based on AVS lateralization benefitted from surgery, including three patients with unilateral unsuccessful AVS and aldosterone suppression in the catheterized side vs inferior vena cava. Radiographically identified adrenal nodules are not always a source of PA, even when ipsilateral with AVS lateralization. These data caution against reliance on imaging findings, either alone or in conjunction with AVS, to guide surgery for PA.
Sigma-Aldrich
Anti-CYP11B2 Antibody, clone 41-17B, clone 41-17B, from mouse