Merck
  • The characterization of alpha5-integrin expression on tubular epithelium during renal injury.
The characterization of alpha5-integrin expression on tubular epithelium during renal injury.
American journal of physiology. Renal physiology (2006-10-05)
Lindsay R White, Jason B Blanchette, Li Ren, Ali Awn, Kiril Trpkov, Daniel A Muruve
ABSTRACT
The hallmark of progressive chronic kidney disease is the deposition of extracellular matrix proteins and tubulointerstitial fibrosis. Integrins mediate cell-extracellular matrix interaction and may play a role tubular epithelial injury. Murine primary tubular epithelial cells (TECs) express alpha(5)-integrin, a fibroblast marker and the natural receptor for fibronectin. Microscopy localized alpha(5)-integrin on E-cadherin-positive cells, confirming epithelial expression. The expression of alpha(5)-integrin increased in TECs grown on fibronectin and occurred in parallel with an upregulation of alpha-smooth muscle actin (alphaSMA), a marker of epithelial-mesenchymal transition (EMT). Exposure of TECs to transforming growth factor (TGF)-beta also increased TEC alpha(5)-integrin expression in association with alphaSMA and EMT. Knock-down of alpha5-integrin expression with short interfering RNA attenuated the TGF-beta induction of alphaSMA but did not alter morphologic EMT. Rather, alpha5-integrin was necessary for epithelial cell migration on fibronectin but not type IV collagen during cell spreading and epithelial wound healing in vitro. Immunohistochemistry revealed basolateral tubular epithelial alpha(5)-integrin expression in mouse kidneys after unilateral ureteric obstruction but not in contralateral control kidneys. In patient biopsies of nondiabetic kidney disease, alpha(5)-integrin expression was increased significantly in the renal interstitium. Focal basolateral staining was also detected in injured, but not in normal, tubular epithelium. In summary, these data show that TECs are induced to express alpha(5)-integrin during EMT and tubular epithelial injury in vitro and in vivo. These results increase our understanding of the biology of integrins during EMT and tubular injury in chronic kidney disease.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-ZO-1 Antibody, clone R40.76, clone R40.76, from rat