Merck
  • SNX9 regulates tubular invagination of the plasma membrane through interaction with actin cytoskeleton and dynamin 2.
SNX9 regulates tubular invagination of the plasma membrane through interaction with actin cytoskeleton and dynamin 2.
Journal of cell science (2008-04-05)
Narae Shin, Namhui Ahn, Belle Chang-Ileto, Joohyun Park, Kohji Takei, Sang-Gun Ahn, Soo-A Kim, Gilbert Di Paolo, Sunghoe Chang
ABSTRACT
Dynamic membrane remodeling during intracellular trafficking is controlled by the intricate interplay between lipids and proteins. BAR domains are modules that participate in endocytic processes by binding and deforming the lipid bilayer. Sorting nexin 9 (SNX9), which functions in clathrin-mediated endocytosis, contains a BAR domain, however, the properties of this domain are not well understood. Here we show that SNX9 shares many properties with other BAR domain-containing proteins, such as amphiphysin and endophilin. SNX9 is able to deform the plasma membrane, as well as liposomes, into narrow tubules and recruit N-WASP and dynamin 2 to these tubules via its SH3 domain. SNX9-induced tubulation is antagonized by N-WASP and dynamin 2 while it is enhanced by perturbation of actin dynamics. However, SNX9 also has several unique properties. The tubulating activity requires the BAR and PX domains, as well as the low-complexity (LC) domain, which binds the Arp2/3 complex. SNX9 also binds to PtdIns(4)P-5-kinases via its PX domain and its tubulating activity is regulated by phosphoinositides. In addition, the kinase activity of PtdIns(4)P-5-kinases is stimulated by interaction with SNX9, suggesting a positive feedback interaction between SNX9 and PtdIns(4)P-5-kinases. These results suggest that SNX9 functions in the coordination of membrane remodeling and fission via interactions with actin-regulating proteins, endocytic proteins and PtdIns(4,5)P2-metabolizing enzymes.
Sigma-Aldrich
Brain Extract from bovine brain, Type I, Folch Fraction I
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphoinositides sodium salt from bovine brain
Pricing and availability is not currently available.