
Triptophenolide

Synonym(s):
(3bR-trans)-3b,4,5,9b,10,11-Hexahydro-6-hydroxy-9b-methyl-7-(1-methylethyl)-phenanthro(1,2-c)furan-1(3H)-one, Hypolide
Empirical Formula (Hill Notation):
C20H24O3
CAS Number:
Molecular Weight:
312.40
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing