
Sodium acetate-d3

Synonym(s):
Acetic-d3 acid sodium salt
Linear Formula:
CD3CO2Na
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing