
Silylating mixture III

Synonym(s):
N,O-Bis-(trimethylsilyl)-trifluoracetamid – chlorotrimethylsilane – 1-(trimethylsilyl)imidazole mixture
Products
Brand
Physical Form
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing