
Sesame Oil Related Compound B

Synonym(s):
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, 1,2-Di(cis-9,12-octadecadienoyl)-3-hexadecanoyl-rac-glycerol, PLL
Empirical Formula (Hill Notation):
C55H98O6
CAS Number:
Molecular Weight:
855.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2