Skip to Content
Merck
(R)-(+)-α-Methylbenzyl isocyanate for chiral derivatization, LiChropur™, ≥99.0%

(R)-(+)-α-Methylbenzyl isocyanate

(R)-(+)-α-Methylbenzyl isocyanate for chiral derivatization, LiChropur™, ≥99.0%
Synonym(s):
(R)-(+)-1-Phenylethyl isocyanate
Linear Formula:
C6H5CH(CH3)NCO
CAS Number:
Molecular Weight:
147.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2