
Punicalagin (A + B mixture)

Synonym(s):
Punicalagin
Empirical Formula (Hill Notation):
C48H28O30
CAS Number:
Molecular Weight:
1084.72
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing