
Prilocaine hydrochloride

Synonym(s):
N-(2-Methylphenyl)-2-(propylamino)propanamide hydrochloride, Prilocaine hydrochloride
Empirical Formula (Hill Notation):
C13H20N2O · HCl
CAS Number:
Molecular Weight:
256.77
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing