
Pregabalin-D6 solution

Empirical Formula (Hill Notation):
C8D6H11NO2
Molecular Weight:
165.26
EC Number:
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2