
Picein

Synonym(s):
1-[4-(β-D-Glucopyranosyloxy)phenyl]ethan-1-one, 4′-(β-D-Glucopyranosyloxy)acetophenone, 4-Acetylphenyl β-D-glucopyranoside, p-Hydroxyacetophenone glucoside, Ameliaroside, Piceoside, Salicinerein, Salinigrin
Empirical Formula (Hill Notation):
C14H18O7
CAS Number:
Molecular Weight:
298.29
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing