
Naproxen Impurity M

Synonym(s):
2-Methoxynaphthalene, Methyl 2-naphthyl ether
Linear Formula:
C10H7OCH3
CAS Number:
Molecular Weight:
158.20
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing