
Monoclonal Anti-FITC−Alkaline Phosphatase antibody produced in mouse

Synonym(s):
Monoclonal Anti-FITC
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Recombinant Host
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing