
Moexipril Related Compound B

Synonym(s):
Moexipril diketopiperazine, (S)-Ethyl 2-{(3S,11aS)-8,9-dimethoxy-3-methyl-1,4-dioxo-3,4-dihydro-1H-pyrazino[1,2-b]isoquinolin-2(6H,11H,11aH)-yl}-4-phenylbutanoate
Empirical Formula (Hill Notation):
C27H32N2O6
CAS Number:
Molecular Weight:
480.55
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2