
Methacrifos

Empirical Formula (Hill Notation):
C7H13O5PS
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing