
Melittin from honey bee venom

Empirical Formula (Hill Notation):
C131H229N39O31
CAS Number:
Molecular Weight:
2846.46
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing