
Licochalcone A

Synonym(s):
(E)-3-[5-(1,1-Dimethyl-2-propenyl)-4-hydroxy-2-methoxyphenyl]-1-(4-hydroxyphenyl)-2-propen-1-one, 4′,4-Dihydroxy-3-α,α-dimethylallyl-6-methoxychalcone
Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing