
Fenthion oxon sulfoxide

Synonym(s):
Dimethyl 3-methyl-4-(methylsulfinyl)phenyl phosphate, Fenoxon sulfoxide
Empirical Formula (Hill Notation):
C10H15O5PS
CAS Number:
Molecular Weight:
278.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2