
Diazinon solution

Empirical Formula (Hill Notation):
C12H21N2O3PS
CAS Number:
Molecular Weight:
304.35
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing