
Anti-MCM3 antibody produced in rabbit

Synonym(s):
Anti-DNA polymerase α holoenzyme-associated protein P1 antibody produced in rabbit, Anti-DNA replication licensing factor MCM3 antibody produced in rabbit, Anti-P1-MCM3 antibody produced in rabbit, Anti-P102 protein antibody produced in rabbit, Anti-RLF subunit β antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing