
Anti-Hepatocyte Growth Factor antibody produced in goat

Synonym(s):
Anti-HGF
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2