
2-Chloro-2′-deoxyadenosine

Synonym(s):
CdA, Cladribine, 2-Chloro-2′-deoxyadenosine
Empirical Formula (Hill Notation):
C10H12ClN5O3
CAS Number:
Molecular Weight:
285.69
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2