2-Bromopropane-d7
Synonym(s):
Isopropyl-d7 bromide
Linear Formula:
(CD3)2CDBr
CAS Number:
Molecular Weight:
130.03
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing