Skip to Content
Merck
All Photos(1)

Documents

61868-U

Proteomix® HIC Ethyl-NP5 (5 µm) HPLC Columns

L × I.D. 10 mm × 4 mm

Synonym(s):

Proteomix® HIC Ethyl-NP5, Guard Cartridge with Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Proteomix® HIC Ethyl-NP5, Guard Cartridge with holder, 5 μm, NP, L × I.D. 10 mm × 4 mm

material

stainless steel hardware

product line

Proteomix®

manufacturer/tradename

Sepax 432NP5-4001C

packaging

1 ea of

parameter

6000 psi max. pressure
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

10 mm × 4 mm

impurities

<10 ppm metals

matrix

polymeric
polystyrene/divinylbenzene (Spherical, highly cross-linked)

matrix active group

ethyl phase

particle size

5 μm

pore size

non-porous

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Analysis Note

Compatible with aqueous solution, a mixture of water and acetonitrile, acetone, methanol, or THF.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service